Learn More
Miele, Inc. APLW 030/1lower basket
Catalog No. 501953665
Encompass_Preferred
APLW 030/1 Lower basket- 12 injectors- Ø max. 100 mm- Height max.: 180-280 mm- incl. nozzles 6x APLW 067, 6x APLW 066
Catalog No. 50-195-3665 Supplier Miele, Inc. Supplier No. 11104890
APLW 030/1 Lower basket- 12 injectors- Ø max. 100 mm- Height max.: 180-280 mm- incl. nozzles 6x APLW 067, 6x APLW 066