Learn More
Miele, Inc. APLW 032/1lower basket
Catalog No. 501953668
Encompass_Preferred
APLW 032/1 Lower basket- 24 + 121 injectors- Ø max. 20 + 75 mm- Height max.: 160-300 mm- incl. nozzles 121x APLW 048, 24x APLW 056
Catalog No. 50-195-3668 Supplier Miele, Inc. Supplier No. 11104920
Add to Cart
Add to Cart
Request a Quote
APLW 032/1 Lower basket- 24 + 121 injectors- Ø max. 20 + 75 mm- Height max.: 160-300 mm- incl. nozzles 121x APLW 048, 24x APLW 056