Learn More
Miele, Inc. APLW 058injector nozzle
Catalog No. 501953694
Encompass_Preferred
APLW 058 Nozzle- A: 6 mm- B: 10 mm- C: 75 mm- D: 135 mm- E: 105 mm
Catalog No. 50-195-3694 Supplier Miele, Inc. Supplier No. 11105180
APLW 058 Nozzle- A: 6 mm- B: 10 mm- C: 75 mm- D: 135 mm- E: 105 mm