Learn More
Miele, Inc. APLW 085bottle neck holder
Catalog No. 501953721
Encompass_Preferred
APLW 085 Bottle neck holder- ø 33 mm- for injectors ø 6 mm
Catalog No. 50-195-3721 Supplier Miele, Inc. Supplier No. 11105450
APLW 085 Bottle neck holder- ø 33 mm- for injectors ø 6 mm