Learn More
Medchemexpress LLC APN-C3-PEG4-azide | 2183440-32-4 | 98.0% | C24H32N6O6 | 25 MG
SDP
Catalog No. 5000352660
Encompass_Preferred
APN-C3-PEG4-azide is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. This click chemistry reagent contains an Azide group, enabling it to participate in copper-catalyzed azide-alkyne cycloaddition (CuAAC) reactions with Alkyne groups or strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Utilized in PROTAC synthesis
  • Click chemistry reagent
  • Contains an azide group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAC)
  • Supports strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Relevant to cancer targeted therapy and PROTAC research
Catalog No. 50-003-52660 Supplier Medchemexpress LLC Supplier No. 25MGHY140841
APN-C3-PEG4-azide is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. This click chemistry reagent contains an Azide group, enabling it to participate in copper-catalyzed azide-alkyne cycloaddition (CuAAC) reactions with Alkyne groups or strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Utilized in PROTAC synthesis
  • Click chemistry reagent
  • Contains an azide group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAC)
  • Supports strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Relevant to cancer targeted therapy and PROTAC research