Learn More
Medchemexpress LLC Apn-C3-peg4-azide | 2183440-32-4 | 98.6% | C24H32N6O6 | 10MG
SDP
Catalog No. 5000260349
Encompass_Preferred
APN-C3-PEG4-azide is a PEG-based heterobifunctional linker for PROTAC assembly and bioconjugation. It combines a thiol-reactive 3-arylpropiolonitrile (APN) group with a PEG4 spacer terminating in an azide, enabling selective cysteine modification and subsequent conjugation via copper-catalyzed or strain-promoted azide-alkyne cycloadditions.
  • Thiol-reactive APN moiety enables selective cysteine conjugation.
  • PEG4 spacer provides flexibility and water solubility.
  • Terminal azide enables copper-catalyzed and strain-promoted click reactions.
  • High purity suitable for synthetic and bioconjugation workflows.
  • Available in multiple small-scale quantities for research use.
Catalog No. 50-002-60349 Supplier Medchemexpress LLC Supplier No. HY14084110MG
APN-C3-PEG4-azide is a PEG-based heterobifunctional linker for PROTAC assembly and bioconjugation. It combines a thiol-reactive 3-arylpropiolonitrile (APN) group with a PEG4 spacer terminating in an azide, enabling selective cysteine modification and subsequent conjugation via copper-catalyzed or strain-promoted azide-alkyne cycloadditions.
  • Thiol-reactive APN moiety enables selective cysteine conjugation.
  • PEG4 spacer provides flexibility and water solubility.
  • Terminal azide enables copper-catalyzed and strain-promoted click reactions.
  • High purity suitable for synthetic and bioconjugation workflows.
  • Available in multiple small-scale quantities for research use.