Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Apn-PEG36-tetrazine | 1997.30 g·mol⁻1 | C94H161N7O38 | 10 MG
SDP

Catalog No. 5000203039
Encompass_Preferred

APN-PEG36-tetrazine is a tetrazine-functionalized polyethylene glycol (PEG36) reagent for bioorthogonal click chemistry and antibody-drug conjugate (ADC) linker development. It undergoes inverse electron-demand Diels-Alder (iEDDA) reactions with trans-cyclooctene (TCO) groups, enabling rapid conjugation in bioconjugation workflows. The reagent is supplied in small research quantities and should be stored protected from light under inert conditions.

  • Tetrazine-functionalized PEG36 for iEDDA click reactions.
  • Suitable for antibody-drug conjugate (ADC) linker synthesis and bioconjugation.
  • Molecular weight approximately 1997.30 g·mol⁻1.
  • Available in small vial sizes for research use (1 mg, 5 mg, 10 mg).
  • Store at -20°C protected from light; in solvent store at -80°C for long-term stability.

Catalog No. 50-002-03039 Supplier Medchemexpress LLC Supplier No. HY13985910MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

APN-PEG36-tetrazine is a tetrazine-functionalized polyethylene glycol (PEG36) reagent for bioorthogonal click chemistry and antibody-drug conjugate (ADC) linker development. It undergoes inverse electron-demand Diels-Alder (iEDDA) reactions with trans-cyclooctene (TCO) groups, enabling rapid conjugation in bioconjugation workflows. The reagent is supplied in small research quantities and should be stored protected from light under inert conditions.

  • Tetrazine-functionalized PEG36 for iEDDA click reactions.
  • Suitable for antibody-drug conjugate (ADC) linker synthesis and bioconjugation.
  • Molecular weight approximately 1997.30 g·mol⁻1.
  • Available in small vial sizes for research use (1 mg, 5 mg, 10 mg).
  • Store at -20°C protected from light; in solvent store at -80°C for long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.