Learn More
Key Scientific Products APNA Gram Reaction Discs, for Best Results, Use Fresh Cultures Less than 48 Hours Old
SDP
Catalog No. NC9503153
Encompass_Preferred
APNA is a rapid test used to confirm gram variable aerobic and facultative anaerobic organisms as gram positive or gram negative.
Catalog No. NC9503153 Supplier Key Scientific Products Supplier No. K915
APNA is a rapid test used to confirm gram variable aerobic and facultative anaerobic organisms as gram positive or gram negative.