Learn More
Key Scientific Products APNA Gram Reaction Discs, 30 per Pack
SDP
Catalog No. NC9560002
Encompass_Preferred
Gram Reaction (APNA) Discs are used in confirming the gram staining reaction of Gram-negative and Gram-positive aerobic and facultatively anaerobic bacteria
Catalog No. NC9560002 Supplier Key Scientific Products Supplier No. K915B
Gram Reaction (APNA) Discs are used in confirming the gram staining reaction of Gram-negative and Gram-positive aerobic and facultatively anaerobic bacteria