Learn More
TARGETMOL CHEMICALS INC APNEA 5MG
SDP
Catalog No. 502581479
Encompass_Preferred
Also available in 10 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. APNEA ((2R3R4S5R)-2-[6-[2-(4-aminophenyl)ethylamino]purin-9-yl]-5-(hydroxymethyl)oxolane-34-diol) is a non-selective agonist of adenosine A3 receptor. purity: 99%
Catalog No. 50-258-1479 Supplier TARGETMOL CHEMICALS INC Supplier No. T121595MG
Also available in 10 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. APNEA ((2R3R4S5R)-2-[6-[2-(4-aminophenyl)ethylamino]purin-9-yl]-5-(hydroxymethyl)oxolane-34-diol) is a non-selective agonist of adenosine A3 receptor. purity: 99%