Learn More
Enzo Life Sciences APO-BRDU™ kit (1Kit)
SDP
Supplier:  Enzo Life Sciences ALX850042KI01
Encompass_Preferred
APO-BRDU™ Kit is a two color TUNEL (Terminal deoxynucleotide transferase dUTP Nick End Labeling) assay for labeling DNA breaks and total cellular DNA to detect apoptotic cells by flow cytometry. The kit contains the instructions and reagents required for measuring apoptosis in cells including: positive and negative control cells for assessing reagent performance; washing, reaction and rinsing buffers for processing individual steps in the assay; terminal deoxynucleotidyl tranferase enzyme (TdT), bromodeoxyuridine triphosphate (Br‐dUTP), and fluorescein labeled anti-BrdU antibody for labeling DNA breaks and propidium iodide/RNase A solution for counter staining the total DNA. Applications: Flow Cytometry .
Catalog No. 50-201-0101