Learn More
Enzo Life Sciences APO-E2 (human), (recombinant) (500µg)
SDP
Catalog No. 502013940
Encompass_Preferred
Alternative name: Apolipoprotein E. Purity: ≥98% (SDS-PAGE gel & HPLC). Formulation: Lyophilized from 20mM sodium phosphate, pH 7.8, containing 0.5 mM DTT. Source: Produced in E.coli. Long Term Storage: -20°C.
Catalog No. 50-201-3940 Supplier Enzo Life Sciences Supplier No. ENZPRT1500500
Alternative name: Apolipoprotein E. Purity: ≥98% (SDS-PAGE gel & HPLC). Formulation: Lyophilized from 20mM sodium phosphate, pH 7.8, containing 0.5 mM DTT. Source: Produced in E.coli. Long Term Storage: -20°C.