Learn More
Medchemexpress LLC ApoA-IV Mouse P.pa 100ug | 100ug
SDP
Catalog No. 5000195818
Encompass_Preferred
ApoA-IV Protein plays a vital role in lipid metabolism influencing chylomicron and VLDL secretion and catabolism It efficiently activates lipoprotein lipase with ApoC-II and serves as a potent activator of LCAT As a component of HDL and chylomicrons ApoA-IV is integral to cholesterol transport and metabolism Structurally the protein forms homodimers reflecting its crucial role in lipid-related processes ApoA-IV Protein Mouse (P pastoris His) is the recombinant mouse-derived ApoA-IV protein expressed by P pastoris with N-6 His labeled tag
Catalog No. 50-001-95818 Supplier Medchemexpress LLC Supplier No. HYP71715100UG
ApoA-IV Protein plays a vital role in lipid metabolism influencing chylomicron and VLDL secretion and catabolism It efficiently activates lipoprotein lipase with ApoC-II and serves as a potent activator of LCAT As a component of HDL and chylomicrons ApoA-IV is integral to cholesterol transport and metabolism Structurally the protein forms homodimers reflecting its crucial role in lipid-related processes ApoA-IV Protein Mouse (P pastoris His) is the recombinant mouse-derived ApoA-IV protein expressed by P pastoris with N-6 His labeled tag