Learn More
Aviva Systems Biology Corp Apoa1 Antibody - N-terminal region (ARP54279_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54279_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Apoa1. It was validated on Western Blot. . Alternate names: apoA-IApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-285-902