Learn More
Biomatik Corporation APOA1 Monoclonal Antibody
Supplier:  Biomatik Corporation CAF50508
Encompass_Preferred
APOA1 Monoclonal Antibody is a Human (Homo sapiens) Monoclonal Antibody to Apolipoprotein A-I. The APOA1 Monoclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Mouse. Species: Human (Homo sapiens). Immunogen: Please Inquire. Applications: ELISA, WB, IHC, IF. Target Synonyms: Apolipoprotein A-I. Accession Number: P02647. Gene ID: 335. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6504