Learn More
Aviva Systems Biology Corp APOA2 Antibody - N-terminal region (ARP54588_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54588_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against APOA2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human APOA2. Alternate names: apoAII, Apo-AII, ApoA-IIApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-62854