Learn More
BioVendor, LLC Apoa2 Human Elisa BioVendor LLC
SDP
Supplier:  BioVendor, LLC REH013R
Encompass
BioVendor Human ApoA2 ELISA kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human ApoA2 in serum, plasma and cell culture supernatants. This assay employs an antibody specific for human ApoA2 coated on a 96-well plate. Standards and samples are pipetted into the wells and ApoA2 present in a sample is bound to the wells by the immobilized antibody. The wells are washed and biotinylated antihuman ApoA2 antibody is added. After washing away unbound biotinylated antibody, HRP conjugated streptavidin is pipetted to the wells. The wells are again washed, a TMB substrate solution is added to the wells and color develops in proportion to the amount of ApoA2 bound. The Stop Solution changes the color from blue to yellow, and the intensity of the color is measured at 450 nm. One kit (96 wells). Calibration range: 0.027-20ng/ml. Limit of detection: 0.025ng/ml. Other names: ApoA2, Apolipoprotein A-II, ApoA-2.
Catalog No. 50-240-6356