Learn More
Medchemexpress LLC Apoatropine hydrochloride | 5978-81-4 | MFCD10574677 | 99.6% | 307.82 g/mol | C17H22ClNO2 | 25 MG
SDP
Catalog No. 5000241670
Encompass_Preferred
Apoatropine hydrochloride is the hydrochloride salt of apoatropine, a tropane alkaloid isolated from Atropa belladonna. It is provided for research use and is used in pharmacological and biochemical studies involving tropane alkaloids, receptor interactions, and analytical characterization.
  • High purity (99.6%) for reliable analytical and experimental results.
  • Molecular weight 307.82 g/mol for accurate formulation and calculations.
  • Hydrochloride salt form improves solubility in polar solvents.
  • Available in small milligram quantities suitable for research and reference use.
  • Intended for research use only and not for diagnostic or therapeutic applications.
Catalog No. 50-002-41670 Supplier Medchemexpress LLC Supplier No. HY105797A25MG
Apoatropine hydrochloride is the hydrochloride salt of apoatropine, a tropane alkaloid isolated from Atropa belladonna. It is provided for research use and is used in pharmacological and biochemical studies involving tropane alkaloids, receptor interactions, and analytical characterization.
  • High purity (99.6%) for reliable analytical and experimental results.
  • Molecular weight 307.82 g/mol for accurate formulation and calculations.
  • Hydrochloride salt form improves solubility in polar solvents.
  • Available in small milligram quantities suitable for research and reference use.
  • Intended for research use only and not for diagnostic or therapeutic applications.