Learn More
BioVendor, LLC Apob-100 Human Elisa BioVendor LLC
SDP
Supplier:  BioVendor, LLC REH004R
Encompass
The BioVendor Human ApoB100 ELISA kit is an in-vitro enzyme-linked immunosorbent assay for the quantitative measurement of human ApoB100 in serum, plasma and cell culture supernatants. This assay employs an antibody specific for human ApoB100 coated on a 96 well plate. Standards and samples are pipetted into the wells and ApoB100 present in a sample is bound to the wells by the immobilized antibody. The wells are washed and biotinylated anti-human ApoB100 antibody is added. After washing away unbound biotinylated antibody, HRP-conjugated streptavidin is pipetted to the wells. The wells are again washed, a TMB substrate solution is added to the wells and color develops in proportion to the amount of ApoB100 bound. The Stop Solution changes the color from blue to yellow, and the intensity of the color is measured at 450 nm. Sample Requirement: 2¿l. Calibration range: 2.458-600ng/ml. Limit of detection: 2.5ng/ml.
Catalog No. 50-240-6354