Learn More
Aviva Systems Biology Corp APOBEC1 Antibody - N-terminal region (ARP40371_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40371_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against APOBEC1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human APOBEC1. Alternate names: APOBEC-1, BEDP, CDAR1, HEPRApplicationWB. Species reactivity: Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-57416