Learn More
New England Biolabs, Inc. APOBEC3A - 800 units
SDP
Catalog No. 5000332615
Encompass
APOBEC3A (Apolipoprotein B mRNA editing enzyme, catalytic polypeptide-like 3A) is a cytidine deaminase enzyme that catalyzes the deamination of cytosine or 5-methylcytosine in single-stranded DNA to produce uracil or thymine.
  • Ideal for cytosine deamination
  • Ideal for APOBEC-Coupled Epigenetic Sequencing (ACE-Seq)
  • Isolated from a recombinant source
  • Supplied with 10X Reaction Buffer
Catalog No. 50-003-32615 Supplier New England Biolabs, Inc. Supplier No. M0648S
APOBEC3A (Apolipoprotein B mRNA editing enzyme, catalytic polypeptide-like 3A) is a cytidine deaminase enzyme that catalyzes the deamination of cytosine or 5-methylcytosine in single-stranded DNA to produce uracil or thymine.
  • Ideal for cytosine deamination
  • Ideal for APOBEC-Coupled Epigenetic Sequencing (ACE-Seq)
  • Isolated from a recombinant source
  • Supplied with 10X Reaction Buffer