Learn More
ABclonal Technology APOBEC3C Rabbit pAb
SDP
Supplier:  ABclonal Technology A14510500UL
Encompass_Preferred
This gene is a member of the cytidine deaminase gene family. It is one of seven related genes or pseudogenes found in a cluster thought to result from gene duplication, on chromosome 22. Members of the cluster encode proteins that are structurally and functionally related to the C to U RNA-editing cytidine deaminase APOBEC1. It is thought that the proteins may be RNA editing enzymes and have roles in growth or cell cycle control.
Catalog No. 50-001-08269