Learn More
Aviva Systems Biology Corp APOBEC3F Antibody - C-terminal region (ARP41136_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41136_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against APOBEC3F. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human APOBEC3F. Alternate names: ARP8, BK150C2.4.MRNA, KA6, MGC74891ApplicationWB. Species reactivity: Human, Pig
Catalog No. 50-014-57733