Learn More
Aviva Systems Biology Corp APOBEC3G Antibody - N-terminal region (ARP33540_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33540_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against APOBEC3G. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human APOBEC3G. Alternate names: ARCD, ARP9, ARP-9, CEM15, CEM-15, MDS019, bK150C2.7, dJ494G10.1ApplicationWB. Species reactivity: Human, Pig
Catalog No. 50-014-55061