Learn More
Aviva Systems Biology Corp APOBEC4 Antibody - N-terminal region (ARP55975_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55975_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against APOBEC4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human APOBEC4. Alternate names: C1orf169, MGC26594ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Pig, Rat
Catalog No. 50-014-61680