Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC APOC3 Human His-SU 10ug
SDP

Catalog No. 5000195577
Encompass_Preferred

Recombinant human apolipoprotein C-III (APOC3) with an N-terminal 6xHis-SUMO fusion, expressed in Escherichia coli and supplied as a lyophilized powder. The protein is highly pure and low in endotoxin, and is formatted and supplied with handling guidance for reliable use in biochemical assays, antibody validation, and binding studies.

  • Contains N-terminal 6xHis and SUMO tags for detection and purification.
  • Derived from human sequence corresponding to residues S21-A99.
  • Expressed in Escherichia coli and provided as a lyophilized powder.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Endotoxin level below 1 EU/μg, suitable for sensitive assays.
  • Includes reconstitution and storage recommendations to preserve activity.
  • Available in small vial sizes for low-volume applications.

Catalog No. 50-001-95577 Supplier Medchemexpress LLC Supplier No. HYP7208110UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human apolipoprotein C-III (APOC3) with an N-terminal 6xHis-SUMO fusion, expressed in Escherichia coli and supplied as a lyophilized powder. The protein is highly pure and low in endotoxin, and is formatted and supplied with handling guidance for reliable use in biochemical assays, antibody validation, and binding studies.

  • Contains N-terminal 6xHis and SUMO tags for detection and purification.
  • Derived from human sequence corresponding to residues S21-A99.
  • Expressed in Escherichia coli and provided as a lyophilized powder.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Endotoxin level below 1 EU/μg, suitable for sensitive assays.
  • Includes reconstitution and storage recommendations to preserve activity.
  • Available in small vial sizes for low-volume applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.