Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC APOC3 Human His-SU 20ug
SDP

Catalog No. 5000195409
Encompass_Preferred

Recombinant human apolipoprotein C-III supplied as a lyophilized His-SUMO-tagged fusion protein for research use. The construct corresponds to UniProt accession P02656 (residues S21-A99), is produced in E. coli, and is offered in microgram-scale sizes. Consult the provided datasheet, COA, and SDS for handling, reconstitution, and storage details.

  • Recombinant human apolipoprotein C-III with N-terminal 6xHis and SUMO tags.
  • Expressed in E. coli for efficient production.
  • Lyophilized powder format for convenient handling and storage.
  • Molecular weight approximately 22-24.8 kDa.
  • Corresponds to UniProt accession P02656 (S21-A99).
  • Stable when stored at -20°C; aliquot and freeze at -20°C or -80°C after reconstitution.
  • Not recommended to reconstitute below 100 μg/mL in ddH2O.

Catalog No. 50-001-95409 Supplier Medchemexpress LLC Supplier No. HYP7208120UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human apolipoprotein C-III supplied as a lyophilized His-SUMO-tagged fusion protein for research use. The construct corresponds to UniProt accession P02656 (residues S21-A99), is produced in E. coli, and is offered in microgram-scale sizes. Consult the provided datasheet, COA, and SDS for handling, reconstitution, and storage details.

  • Recombinant human apolipoprotein C-III with N-terminal 6xHis and SUMO tags.
  • Expressed in E. coli for efficient production.
  • Lyophilized powder format for convenient handling and storage.
  • Molecular weight approximately 22-24.8 kDa.
  • Corresponds to UniProt accession P02656 (S21-A99).
  • Stable when stored at -20°C; aliquot and freeze at -20°C or -80°C after reconstitution.
  • Not recommended to reconstitute below 100 μg/mL in ddH2O.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.