Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC APOC3 Human His-SU 50ug
SDP

Catalog No. 5000195538
Encompass_Preferred

Recombinant human apolipoprotein C-III (mature residues S21-A99) expressed in E. coli and supplied as a lyophilized His-SUMO-tagged protein for research use. The product is characterized by high purity, low endotoxin, and includes recommended reconstitution and storage conditions.

  • Contains N-terminal 6xHis and SUMO tags for affinity purification and improved solubility.
  • Lyophilized powder formulation for long-term storage and convenient handling.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Endotoxin level below 1 EU/μg suitable for biochemical and binding assays.
  • Stable when stored at -20°C; aliquot and freeze at -20°C or -80°C for extended storage.
  • Reconstitute to at least 100 μg/mL; avoid preparing dilute solutions below advised concentration.

Catalog No. 50-001-95538 Supplier Medchemexpress LLC Supplier No. HYP7208150UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human apolipoprotein C-III (mature residues S21-A99) expressed in E. coli and supplied as a lyophilized His-SUMO-tagged protein for research use. The product is characterized by high purity, low endotoxin, and includes recommended reconstitution and storage conditions.

  • Contains N-terminal 6xHis and SUMO tags for affinity purification and improved solubility.
  • Lyophilized powder formulation for long-term storage and convenient handling.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Endotoxin level below 1 EU/μg suitable for biochemical and binding assays.
  • Stable when stored at -20°C; aliquot and freeze at -20°C or -80°C for extended storage.
  • Reconstitute to at least 100 μg/mL; avoid preparing dilute solutions below advised concentration.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.