Learn More
Medchemexpress LLC APOC3 Human His-SU 5ug
SDP
Catalog No. 5000196278
Encompass_Preferred
Recombinant human apolipoprotein C-III (APOC3) produced in E. coli with N-terminal 6xHis and SUMO tags. Supplied as a purified protein in a 5 μg format, it is intended for biochemical assays, antibody validation, binding studies, and functional research into triglyceride metabolism and lipoprotein assembly.
  • Expressed in E. coli with N-terminal 6xHis and SUMO tags for easy detection and purification.
  • Purified recombinant protein suitable for biochemical and binding assays.
  • Compact 5 μg size ideal for pilot experiments and antibody validation.
  • Human sequence suitable for human-relevant functional studies.
  • Provided with documentation including accession information for experimental reproducibility.
Catalog No. 50-001-96278 Supplier Medchemexpress LLC Supplier No. HYP720815UG
Recombinant human apolipoprotein C-III (APOC3) produced in E. coli with N-terminal 6xHis and SUMO tags. Supplied as a purified protein in a 5 μg format, it is intended for biochemical assays, antibody validation, binding studies, and functional research into triglyceride metabolism and lipoprotein assembly.
  • Expressed in E. coli with N-terminal 6xHis and SUMO tags for easy detection and purification.
  • Purified recombinant protein suitable for biochemical and binding assays.
  • Compact 5 μg size ideal for pilot experiments and antibody validation.
  • Human sequence suitable for human-relevant functional studies.
  • Provided with documentation including accession information for experimental reproducibility.