Learn More
Cayman Chemical Apocarotenal, Purity: ≥95%, CAS: 1107-26-2, 100 mg
SDP
Catalog No. 502919377
Encompass_Preferred
A carotenoid with genotoxic and antioxidant activities and an active metabolite of β-carotene; scavenges free radicals in a TEAC assay at 100 µM; increases the percentage of micronucleated cells and the number of sister chromatid exchanges and chromosomal aberrations in primary rat hepatocytes at 10 µM
Catalog No. 50-291-9377 Supplier Cayman Chemical Supplier No. 37231100 MG
A carotenoid with genotoxic and antioxidant activities and an active metabolite of β-carotene; scavenges free radicals in a TEAC assay at 100 µM; increases the percentage of micronucleated cells and the number of sister chromatid exchanges and chromosomal aberrations in primary rat hepatocytes at 10 µM