Learn More
Medchemexpress LLC Apocynin-d3 (Acetovanillone-d3) | 80404-23-5 | 99.9% | 169.19 | 1 MG
SDP
Catalog No. 5000427689
Encompass_Preferred
Apocynin-d3 is the deuterium-labeled form of Apocynin, a selective NADPH-oxidase inhibitor with an IC50 of 10 μM. This compound can be used for various applications:
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-27689 Supplier Medchemexpress LLC Supplier No. HYN0088S1MG
Apocynin-d3 is the deuterium-labeled form of Apocynin, a selective NADPH-oxidase inhibitor with an IC50 of 10 μM. This compound can be used for various applications:
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.