Learn More
Cell Signaling Technology ApoE2 Matched Antibody Pair, Human, Unconjugated, 1 Kit
SDP
Catalog No. 503133918
Encompass
The ApoE2 Matched Antibody Pair is designed for use in immunoassay technologies and high-throughput platforms, such as ELISA, that require antibody pairs suitable for custom or specialized labeling, including biotin, fluorophores, beads, and lanthanides. This pair consists of capture and detection antibodies that bind to non-overlapping epitopes and are supplied in a BSA and azide-free formulation, making them compatible with various conjugation chemistries. The kit is optimized for detecting endogenous protein expression in human cerebrospinal fluid, plasma, and serum samples.
  • Compatible with platforms including MSD, Quanterix Simoa, Luminex, and Alpha technology (e.g., AlphaScreen, AlphaLISA, HTRF)
  • Includes capture and detection antibodies that bind to non-overlapping epitopes
  • Supplied in a BSA and azide-free formulation
  • Validated for the detection of endogenous protein in human samples
Catalog No. 50-313-3918 Supplier Cell Signaling Technology Supplier No. 91181S
The ApoE2 Matched Antibody Pair is designed for use in immunoassay technologies and high-throughput platforms, such as ELISA, that require antibody pairs suitable for custom or specialized labeling, including biotin, fluorophores, beads, and lanthanides. This pair consists of capture and detection antibodies that bind to non-overlapping epitopes and are supplied in a BSA and azide-free formulation, making them compatible with various conjugation chemistries. The kit is optimized for detecting endogenous protein expression in human cerebrospinal fluid, plasma, and serum samples.
  • Compatible with platforms including MSD, Quanterix Simoa, Luminex, and Alpha technology (e.g., AlphaScreen, AlphaLISA, HTRF)
  • Includes capture and detection antibodies that bind to non-overlapping epitopes
  • Supplied in a BSA and azide-free formulation
  • Validated for the detection of endogenous protein in human samples