Learn More
Cayman Chemical ApoE2humnrecombInt 100ug
SDP
Catalog No. 502915428
Encompass_Preferred
Source: Active recombinant human C-terminal His-tagged ApoE2 expressed in HEK293 cells • Amino acids: 19-317 • MW: 37.6 kDa
Catalog No. 50-291-5428 Supplier Cayman Chemical Supplier No. 37225100 UG
Source: Active recombinant human C-terminal His-tagged ApoE2 expressed in HEK293 cells • Amino acids: 19-317 • MW: 37.6 kDa