Learn More
Genscript Corporation APOE3/Apolipoprotein EBiotin
Catalog No. 5000058597
Encompass
Apolipoprotein E (apoE) is a lipid carrier in both the peripheral and the central nervous systems Lipid-loaded apoE lipoprotein particles bind to several cell surface receptors to support membrane homeostasis and injury repair in the brain Considering prevalence and relative risk magnitude the 4 allele of the APOE gene is the strongest genetic risk factor for late-onset Alzheimer s disease (AD)
Catalog No. 50-000-58597 Supplier Genscript Corporation Supplier No. Z04400100
Apolipoprotein E (apoE) is a lipid carrier in both the peripheral and the central nervous systems Lipid-loaded apoE lipoprotein particles bind to several cell surface receptors to support membrane homeostasis and injury repair in the brain Considering prevalence and relative risk magnitude the 4 allele of the APOE gene is the strongest genetic risk factor for late-onset Alzheimer s disease (AD)