Learn More
Cayman Chemical ApoE3humnrecombInt 100ug
SDP
Catalog No. 502915365
Encompass_Preferred
Source: Active recombinant human C-terminal His-tagged ApoE3 expressed in HEK293 cells • Amino acids: 19-317 • MW: 37.7 kDa
Catalog No. 50-291-5365 Supplier Cayman Chemical Supplier No. 37226100 UG
Source: Active recombinant human C-terminal His-tagged ApoE3 expressed in HEK293 cells • Amino acids: 19-317 • MW: 37.7 kDa