Learn More
Enzo Life Sciences ApoE4 monoclonal antibody (4E4) (100µg)
SDP
Supplier:  Enzo Life Sciences ENZABS2950100
Encompass_Preferred
Clone: 4E4. Isotype: IgG1κ. Immunogen: Synthetic peptide corresponding to residues surrounding Arg110 of human ApoE4 protein. Source: Purified from ascites. Species reactivity: Human. Specificity: Recognizes endogenous levels of total ApoE4 protein. Applications: ELISA, IP, WB. Purity: Protein G affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide.
Catalog No. 50-201-2835