Learn More
Cayman Chemical ApoE4humnrecombInt 100ug
SDP
Catalog No. 502919378
Encompass_Preferred
Source: Active recombinant human C-terminal His-tagged ApoE4 expressed in HEK293 cells • Amino acids: 19-317 • MW: 37.8 kDa
Catalog No. 50-291-9378 Supplier Cayman Chemical Supplier No. 37227100 UG
Source: Active recombinant human C-terminal His-tagged ApoE4 expressed in HEK293 cells • Amino acids: 19-317 • MW: 37.8 kDa