Learn More
Aviva Systems Biology Corp Apof Antibody - C-terminal region (ARP41961_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41961_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Apof. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Rat. Alternate names: ApofApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-284-968