Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Apoferritin, from Horse Spleen

Catalog No. AAJ60630MC
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60630MC 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ60630MC Supplier Thermo Scientific Chemicals Supplier No. J60630MC
Add to Cart

missing translation for 'validMainframeMsg'

 

Apoferritin is a gel filtration molecular weight marker that can be used in gel filtration chromatography and protein chromatography. Apoferritin has the ability to demetallate hemin and to metallate protoporphyrin. Apoferritin is used as a novel nanocarrier to load methylene blue (MB) and demonstrate its potential applications for photodynamic therapy of cancer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Molecular Weight Marker - Gel Filtration
Formulation Lyophilized powder, Reconstitute in 0.15M NaCl
Quantity 100 mg
Source Horse spleen
Storage Requirements 2° to 8°C
Regulatory Status RUO
Product Type Apoferritin, from Horse Spleen
Recombinant Native
Form Lyophilized

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.