Learn More
Aviva Systems Biology Corp APOH Antibody - N-terminal region (ARP51158_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51158_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against APOH. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human APOH. Alternate names: B2G1, BG, B2GP1ApplicationWB, IHC. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-60502