Learn More
Aviva Systems Biology Corp APOL2 Antibody - middle region (ARP49974_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49974_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against APOL2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human APOL2. Alternate names: APOL-II, APOL3ApplicationWB. Species reactivity: Human, Rabbit
Catalog No. 50-285-475