Learn More
Aviva Systems Biology Corp APOL6 Antibody - C-terminal region (ARP39649_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39649_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against APOL6. It was validated on Western Blot. . Alternate names: APOL-VI, APOLVI, DKFZp667M075, FLJ38562, FLJ90164, MGC57495ApplicationWB. Species reactivity: Human
Catalog No. 50-284-818