Learn More
Crystal Chem Inc Apolipoprotein A-1 (ApoA1) ELISA Kit, 96-wells
SDP
Supplier:  Crystal Chem Inc 80702
Encompass
The Apolipoprotein A-1 ELISA is used to quantify levels of human apolipoprotein A-1 (ApoA1) in serum and plasma using only a 2 µL sample volume. Highlights: Kit uses sample size of only 2 µL. High sensitivity of 4.1 ng/mL. Dynamic range of 15.6 - 500 ng/mL. Crystal Chem is a worldwide leading supplier of ELISA kits for human markers. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7828