Learn More
Medchemexpress LLC Apolipoprotein A-I | 96.9% | 5 UG
SDP
Catalog No. 5000291427
Encompass_Preferred
Recombinant mouse apolipoprotein A-I (APOA1) produced in HEK293 cells with a C-terminal His tag. The protein is supplied as a 0.22 μm filtered solution in PBS (pH 7.4) containing 5% trehalose, 5% mannitol, and 0.01% Tween 80, and is intended for research use only.
Catalog No. 50-002-91427 Supplier Medchemexpress LLC Supplier No. HYP755035UG
Recombinant mouse apolipoprotein A-I (APOA1) produced in HEK293 cells with a C-terminal His tag. The protein is supplied as a 0.22 μm filtered solution in PBS (pH 7.4) containing 5% trehalose, 5% mannitol, and 0.01% Tween 80, and is intended for research use only.