Learn More
Medchemexpress LLC Apolipoprotein A-I/A 10ug | 10ug
SDP
Catalog No. 5000193676
Encompass_Preferred
The apolipoprotein AI (APOA1) protein is key to reverse cholesterol transport promoting tissue efflux and serves as an important cofactor for lecithin cholesterol acyltransferase (LCAT) promoting cholesterol excretion As a homodimer APOA1 is a component of the sperm activating protein complex (SPAP) interacts with APOA1BP CLU NDRG1 SCGB3A2 NAXE and YJEFN3 exhibiting multiple molecular associations Apolipoprotein A-I/APOA1 Protein Mouse (Truncated HEK293 Fc) is the recombinant mouse-derived Apolipoprotein A-I/APOA1 protein expressed by HEK293 with C-hFc labeled tag
Catalog No. 50-001-93676 Supplier Medchemexpress LLC Supplier No. HYP7823810UG
The apolipoprotein AI (APOA1) protein is key to reverse cholesterol transport promoting tissue efflux and serves as an important cofactor for lecithin cholesterol acyltransferase (LCAT) promoting cholesterol excretion As a homodimer APOA1 is a component of the sperm activating protein complex (SPAP) interacts with APOA1BP CLU NDRG1 SCGB3A2 NAXE and YJEFN3 exhibiting multiple molecular associations Apolipoprotein A-I/APOA1 Protein Mouse (Truncated HEK293 Fc) is the recombinant mouse-derived Apolipoprotein A-I/APOA1 protein expressed by HEK293 with C-hFc labeled tag