Learn More
Medchemexpress LLC Apolipoprotein A-I/A 10ug
SDP
Catalog No. 5000181786
Encompass_Preferred
Apolipoprotein AI plays a key role in reverse cholesterol transport promoting cholesterol efflux from tissues and acting as a cofactor for lecithin cholesterol acyltransferase (LCAT) It forms homodimers and participates in the SPAP complex which is essential for sperm motility interacting with both NAXE and CLU Apolipoprotein A-I/APOA1 Protein Human (HEK293 Fc) is the recombinant human-derived Apolipoprotein A-I/APOA1 protein expressed by HEK293 with C-hFc labeled tag
Catalog No. 50-001-81786 Supplier Medchemexpress LLC Supplier No. HYP7283210UG
Add to Cart
Add to Cart
Apolipoprotein AI plays a key role in reverse cholesterol transport promoting cholesterol efflux from tissues and acting as a cofactor for lecithin cholesterol acyltransferase (LCAT) It forms homodimers and participates in the SPAP complex which is essential for sperm motility interacting with both NAXE and CLU Apolipoprotein A-I/APOA1 Protein Human (HEK293 Fc) is the recombinant human-derived Apolipoprotein A-I/APOA1 protein expressed by HEK293 with C-hFc labeled tag