Learn More
Medchemexpress LLC Apolipoprotein A-I/A 500ug
SDP
Catalog No. 5000181801
Encompass_Preferred
Apolipoprotein AI plays a key role in reverse cholesterol transport promoting cholesterol efflux from tissues and acting as a cofactor for lecithin cholesterol acyltransferase (LCAT) It forms homodimers and participates in the SPAP complex which is essential for sperm motility interacting with both NAXE and CLU Apolipoprotein A-I/APOA1 Protein Human (HEK293 Fc) is the recombinant human-derived Apolipoprotein A-I/APOA1 protein expressed by HEK293 with C-hFc labeled tag
Catalog No. 50-001-81801 Supplier Medchemexpress LLC Supplier No. HYP72832500UG
Apolipoprotein AI plays a key role in reverse cholesterol transport promoting cholesterol efflux from tissues and acting as a cofactor for lecithin cholesterol acyltransferase (LCAT) It forms homodimers and participates in the SPAP complex which is essential for sperm motility interacting with both NAXE and CLU Apolipoprotein A-I/APOA1 Protein Human (HEK293 Fc) is the recombinant human-derived Apolipoprotein A-I/APOA1 protein expressed by HEK293 with C-hFc labeled tag