Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Apolipoprotein A-I (APOA1) | 1426185-50-3 | >95.0% | 100 UG
SDP

Catalog No. 5000292675
Encompass_Preferred

Recombinant human apolipoprotein A-I (His-tag) produced in HEK293 cells. Supplied lyophilized, this purified protein supports biochemical assays, structural studies, and antibody production, and is used to study lipid metabolism and reverse cholesterol transport.

  • Recombinant human protein expressed in HEK293 cells.
  • C-terminal His tag for purification.
  • Lyophilized formulation for stability.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Approximate molecular weight 29 kDa.
  • Available in 100 μg size.
  • Store at -20°C; reconstituted material stable short term at 4°C.

Catalog No. 50-002-92675 Supplier Medchemexpress LLC Supplier No. HYP7525100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human apolipoprotein A-I (His-tag) produced in HEK293 cells. Supplied lyophilized, this purified protein supports biochemical assays, structural studies, and antibody production, and is used to study lipid metabolism and reverse cholesterol transport.

  • Recombinant human protein expressed in HEK293 cells.
  • C-terminal His tag for purification.
  • Lyophilized formulation for stability.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Approximate molecular weight 29 kDa.
  • Available in 100 μg size.
  • Store at -20°C; reconstituted material stable short term at 4°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.