Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Apolipoprotein A-I (APOA1) | 96.9% | 50 UG
SDP

Catalog No. 5000293339
Encompass_Preferred

Recombinant mouse apolipoprotein A-I (APOA1) expressed in HEK293 cells and purified with a C-terminal His tag. Supplied as a 0.22 μm filtered PBS solution (pH 7.4) stabilized for handling, this protein is intended for research applications in lipid metabolism, HDL structure, and cholesterol efflux studies. Store frozen and avoid repeated freeze-thaw cycles.

  • Mouse sequence, HEK293 expression system.
  • C-terminal His tag for detection and purification.
  • Supplied as sterile-filtered solution in PBS, pH 7.4.
  • Stabilized with 5% trehalose and 5% mannitol, 0.01% Tween 80.
  • Low endotoxin level, <1 EU/μg.
  • Molecular weight 27-31 kDa.
  • Supplied quantity 50 UG.
  • For research use only; not for clinical or diagnostic use.

Catalog No. 50-002-93339 Supplier Medchemexpress LLC Supplier No. HYP7550350UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant mouse apolipoprotein A-I (APOA1) expressed in HEK293 cells and purified with a C-terminal His tag. Supplied as a 0.22 μm filtered PBS solution (pH 7.4) stabilized for handling, this protein is intended for research applications in lipid metabolism, HDL structure, and cholesterol efflux studies. Store frozen and avoid repeated freeze-thaw cycles.

  • Mouse sequence, HEK293 expression system.
  • C-terminal His tag for detection and purification.
  • Supplied as sterile-filtered solution in PBS, pH 7.4.
  • Stabilized with 5% trehalose and 5% mannitol, 0.01% Tween 80.
  • Low endotoxin level, <1 EU/μg.
  • Molecular weight 27-31 kDa.
  • Supplied quantity 50 UG.
  • For research use only; not for clinical or diagnostic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.