Learn More
Medchemexpress LLC Apolipoprotein A-I (APOA1) protein, mouse | >95.0% | 50 UG
SDP
Catalog No. 5000291209
Encompass_Preferred
Recombinant mouse apolipoprotein A-I is supplied as a C-terminal 6xHis-tagged, lyophilized protein expressed in HEK293 cells. It is intended for biochemical and cell-based studies of cholesterol efflux, lipid interactions, and protein-protein binding, and is formulated for stability, easy handling, and long-term storage.
  • Mammalian expression preserves native folding and post-translational modifications.
  • High purity (>95.0%) verified by reducing SDS-PAGE.
  • C-terminal His tag facilitates purification and detection.
  • Lyophilized formulation enables extended shelf life and convenient transport.
  • Stable after reconstitution at 4 °C for one week or frozen for long-term storage.
  • Suitable for studies of cholesterol transport, lipid binding, and interaction assays.
Catalog No. 50-002-91209 Supplier Medchemexpress LLC Supplier No. HYP75503A50UG
Recombinant mouse apolipoprotein A-I is supplied as a C-terminal 6xHis-tagged, lyophilized protein expressed in HEK293 cells. It is intended for biochemical and cell-based studies of cholesterol efflux, lipid interactions, and protein-protein binding, and is formulated for stability, easy handling, and long-term storage.
  • Mammalian expression preserves native folding and post-translational modifications.
  • High purity (>95.0%) verified by reducing SDS-PAGE.
  • C-terminal His tag facilitates purification and detection.
  • Lyophilized formulation enables extended shelf life and convenient transport.
  • Stable after reconstitution at 4 °C for one week or frozen for long-term storage.
  • Suitable for studies of cholesterol transport, lipid binding, and interaction assays.